Assay Details
Target Gene Details
Entrez Gene ID: | 67388 |
Gene Name: | RIKEN cDNA 1110008F13 gene |
Gene Aliases: |
AI316789 |
Location: |
Chr.2:156863122-156873563 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008F13Rik | NM_026124.3 | 1 | 48 | NP_080400.1 |
| AK002452.1 | BAB22111.1 | |||
| AK003568.1 | BAB22864.1 | |||
| AK031002.1 | ||||
| AK078178.1 | ||||
| AK155559.1 | ||||
| BC019445.1 | AAH19445.1 | |||
| BY165552.1 | 1 | 48 |

Genomic Map