Assay Details
Target Gene Details
Entrez Gene ID: | 56846 |
Gene Name: | N-terminal EF-hand calcium binding protein 3 |
Gene Aliases: |
2900010M17Rik, AI853434, Apba2bp, Nip1, XB51 |
Location: |
Chr.2:154544399-154559406 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Necab3 | NM_021546.3 | 13 | 1684 | NP_067521.2 |
AB039948.1 | 11 | 1505 | BAB16414.1 | |
AK013520.1 | 12 | 1611 | BAB28895.1 | |
AK044296.1 | 13 | 1676 | BAC31859.1 | |
AK158225.1 | 13 | 1686 | BAE34411.1 |
