Assay Details
Target Gene Details
Entrez Gene ID: | 20181 |
Gene Name: | retinoid X receptor alpha |
Gene Aliases: |
9530071D11Rik, Nr2b1, RXRalpha1 |
Location: |
Chr.2:27676324-27763319 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rxra | NM_001290481.1 | 10 | 2564 | NP_001277410.1 |
| NM_001290482.1 | 10 | 2601 | NP_001277411.1 | |
| NM_011305.3 | 10 | 2743 | NP_035435.1 | |
| AK029121.1 | 1 | 902 | ||
| AK031034.1 | 10 | 2612 | ||
| AK144675.1 | 10 | 2745 | BAE26004.1 | |
| BC138800.1 | 11 | 2594 | ||
| BC138802.1 | 11 | 2594 |
More Information
Set Membership: |
|

Genomic Map