Assay Details
Target Gene Details
Entrez Gene ID: | 66328 |
Gene Name: | SCP2 sterol-binding domain containing 1 |
Gene Aliases: |
1700010M22Rik |
Location: |
Chr.2:144823666-144824415 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scp2d1 | NM_025490.2 | 1 | 633 | NP_079766.3 |
| AK005847.1 | 1 | 634 | BAB24272.2 | |
| AK015138.1 | ||||
| BC048653.1 | AAH48653.1 |
More Information
Set Membership: |
|

Genomic Map