Assay Details
Target Gene Details
Entrez Gene ID: | 73884 |
Gene Name: | zinc finger, DBF-type containing 2 |
Gene Aliases: |
4930431J08Rik, 9330107J05Rik |
Location: |
Chr.1:63200358-63314576 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zdbf2 | NM_001267872.1 | 6 | 1352 | NP_001254801.1 |
NM_001285936.1 | 7 | 1486 | NP_001272865.1 | |
NM_001285937.1 | 6 | 1429 | NP_001272866.1 | |
NM_028673.1 | 7 | 1402 | NP_082949.1 | |
AB777269.1 | 6 | 1429 | ||
AB777270.1 | 7 | 1486 |
