Assay Details
Target Gene Details
Entrez Gene ID: | 228413 |
Gene Name: | proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane) |
Gene Aliases: |
9930111I18Rik, TMG4 |
Location: |
Chr.2:104830741-104849850 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrg4 | NM_178695.5 | 6 | 1118 | NP_848810.3 |
| AK035530.1 | 6 | 943 | BAC29092.1 | |
| AK037087.1 | 6 | 1038 | BAC29697.1 | |
| AK038703.1 | BAC30106.1 | |||
| AK166405.1 | 5 | 1079 | BAE38757.1 | |
| BC099390.1 | AAH99390.1 | |||
| BC115470.2 | ||||
| BC151011.1 | 6 | 1048 | ||
| BC151019.1 | 6 | 1048 |
More Information
Set Membership: |
|

Genomic Map