Assay Details
Target Gene Details
Entrez Gene ID: | 11538 |
Gene Name: | activity-dependent neuroprotective protein |
Gene Aliases: |
AA589558, mKIAA0784 |
Location: |
Chr.2:168180965-168207062 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adnp | NM_009628.2 | 4 | 4202 | NP_033758.2 |
AK004270.1 | 1 | 373 | ||
AK079160.1 | 1 | 899 | BAC37563.1 | |
AK129214.1 | 4 | 4253 | BAC98024.1 | |
AK134786.1 | 3 | 3724 | BAE22284.1 | |
BC050833.1 | 1 | 3151 | AAH50833.1 | |
BC090840.1 | 1 | 3398 | AAH90840.1 |
