Assay Details
Target Gene Details
Entrez Gene ID: | 54485 |
Gene Name: | delta-like 4 (Drosophila) |
Gene Aliases: |
Delta4 |
Location: |
Chr.2:119325784-119335666 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dll4 | NM_019454.3 | 1 | 278 | NP_062327.2 |
| AF253469.1 | 1 | 233 | AAF76428.1 | |
| AF273454.1 | 1 | 299 | AAF78785.1 | |
| AK004739.1 | 1 | 246 | BAB23520.1 | |
| AK031699.1 | 1 | 247 | ||
| AK052322.1 | 1 | 279 | ||
| AK165517.1 | 1 | 247 | ||
| AK171717.1 | 1 | 247 | ||
| BC042497.1 | 1 | 233 | AAH42497.1 | |
| BC049130.1 | 1 | 238 | AAH49130.1 |
Target Gene Details
Entrez Gene ID: | 100043609 |
Gene Name: | predicted gene 14207 |
Gene Aliases: |
Dll4, OTTMUSG00000015946 |
Location: |
Chr.2:119321199-119326197 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm14207 | NR_030683.1 | 1 | 137 | |
| AK039958.1 | 1 | 138 | BAE20559.1 |

Genomic Map