Assay Details
Target Gene Details
Entrez Gene ID: | 629747 |
Gene Name: | serine peptidase inhibitor, Kunitz type, 3 |
Gene Aliases: |
Gm14315, OTTMUSG00000016288 |
Location: |
Chr.2:164569695-164573456 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spint3 | NM_001177401.1 | 2 | 399 | NP_001170872.1 |
| AK136730.1 | 2 | 377 | BAE23110.1 | |
| BU938823.1 | 2 | 402 |
More Information
Set Membership: |
|

Genomic Map