Assay Details
Target Gene Details
Entrez Gene ID: | 338535 |
Gene Name: | RIKEN cDNA E030013I19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:12301246-12312315 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E030013I19Rik | NR_040353.1 | |||
| AK086420.1 | ||||
| AK086944.1 | 1 | 661 | ||
| AK142619.1 |
Target Gene Details
Entrez Gene ID: | 241226 |
Gene Name: | integrin alpha 8 |
Gene Aliases: |
AI447669 |
Location: |
Chr.2:12106632-12301996 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga8 | NM_001001309.2 | 1 | 237 | NP_001001309.1 |
| AF041409.1 | AAC15665.1 | |||
| AK044910.1 | 1 | 239 | BAC32137.1 | |
| AK135193.1 | 1 | 216 | BAE22455.1 |
More Information
Set Membership: |
|

Genomic Map