Assay Details
Target Gene Details
Entrez Gene ID: | 26428 |
Gene Name: | origin recognition complex, subunit 4 |
Gene Aliases: |
Orc4P, Orc4l, mMmORC4 |
Location: |
Chr.2:48902754-48950272 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orc4 | NM_011958.3 | 14 | 2653 | NP_036088.3 |
| NR_033442.1 | 14 | 2601 | ||
| AK084326.1 | 14 | 2622 | BAC39161.1 | |
| AK169291.1 | BAE41048.1 |
More Information
Set Membership: |
|

Genomic Map