Assay Details
Target Gene Details
Entrez Gene ID: | 72080 |
Gene Name: | suppressor APC domain containing 2 |
Gene Aliases: |
2010317E24Rik, 6030458L21Rik, AL033337, ang |
Location: |
Chr.2:25372035-25378213 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sapcd2 | NM_001081085.2 | 1 | 626 | NP_001074554.1 |
NM_001290407.1 | 1 | 626 | NP_001277336.1 | |
AB197927.1 | 1 | 626 | BAD86554.1 | |
AK008577.1 | 1 | 323 | BAB25756.1 | |
AK077952.1 | 1 | 284 | ||
AK154143.1 | 1 | 333 | ||
AK164615.1 | 1 | 301 | BAE37847.1 | |
AK166625.1 | 1 | 301 | BAE38903.1 | |
BC070477.1 | 1 | 327 | AAH70477.1 |
