Assay Details
Target Gene Details
Entrez Gene ID: | 72080 |
Gene Name: | suppressor APC domain containing 2 |
Gene Aliases: |
2010317E24Rik, 6030458L21Rik, AL033337, ang |
Location: |
Chr.2:25372035-25378213 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sapcd2 | NM_001081085.2 | 7 | 2166 | NP_001074554.1 |
NM_001290407.1 | 6 | 2064 | NP_001277336.1 | |
AB197927.1 | 6 | 2064 | BAD86554.1 | |
AK154143.1 | 7 | 1877 | ||
AK163973.1 | 7 | 1385 | BAE37561.1 | |
AK164615.1 | 8 | 1626 | BAE37847.1 | |
AK166625.1 | 7 | 1841 | BAE38903.1 |
