Assay Details
Target Gene Details
Entrez Gene ID: | 13002 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 5 |
Gene Aliases: |
2610314I24Rik, AU018536, Csp |
Location: |
Chr.2:181520305-181552886 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc5 | NM_001271584.1 | 6 | 2260 | NP_001258513.1 |
| NM_001271585.1 | 6 | 2287 | NP_001258514.1 | |
| NM_016775.3 | 5 | 2207 | NP_058055.1 | |
| NR_073369.1 | 6 | 2282 | ||
| AK012029.1 | 1 | 529 | ||
| AK029006.1 | 5 | 2145 | BAC26236.1 | |
| AK032373.1 | 5 | 2139 | BAC27841.1 | |
| AK083584.1 | 6 | 2244 | ||
| AK179639.1 | 1 | 195 | ||
| BC012268.1 | 6 | 2177 | AAH12268.1 | |
| BC024465.1 | 1 | 297 | AAH24465.1 |
More Information
Set Membership: |
|

Genomic Map