Assay Details
Target Gene Details
Entrez Gene ID: | 14231 |
Gene Name: | FK506 binding protein 7 |
Gene Aliases: |
23kDa, FKBP-7, FKBP23 |
Location: |
Chr.2:76663034-76673139 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp7 | NM_010222.2 | 1 | 198 | NP_034352.1 |
| AF040252.1 | 1 | 170 | AAC79959.1 | |
| AK042208.1 | 1 | 186 | ||
| AK138924.1 | 1 | 114 | BAE23818.1 | |
| BC032961.1 | 1 | 155 | AAH32961.1 |
More Information
Set Membership: |
|

Genomic Map