Assay Details
Target Gene Details
Entrez Gene ID: | 99152 |
Gene Name: | anaphase promoting complex subunit 2 |
Gene Aliases: |
9230107K09Rik, AL024279, Apc2, Emi4, Imi4, mKIAA1406 |
Location: |
Chr.2:25272466-25285916 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc2 | NM_175300.4 | 2 | 317 | NP_780509.2 |
| AK033784.1 | 2 | 285 | BAC28472.1 | |
| AK129351.1 | 2 | 270 | BAC98161.1 | |
| AK154465.1 | 2 | 271 | BAE32605.1 | |
| AK170676.1 | 2 | 283 | BAE41951.1 | |
| BC027351.1 | 2 | 265 | AAH27351.1 | |
| BC137583.1 | 2 | 285 | ||
| CJ072758.1 | 2 | 318 | ||
| CJ164053.1 | 2 | 317 |

Genomic Map