Assay Details
Target Gene Details
Entrez Gene ID: | 12848 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 2 (Arabidopsis thaliana) |
Gene Aliases: |
AI315723, C85265, Csn2, Sgn2, TRIP-15, Trip15 |
Location: |
Chr.2:125830302-125859082 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops2 | NM_001285507.1 | 13 | 1986 | NP_001272436.1 |
| NM_001285512.1 | 13 | 2013 | NP_001272441.1 | |
| NM_001285513.1 | 12 | 1777 | NP_001272442.1 | |
| NM_009939.3 | 13 | 1965 | NP_034069.2 | |
| AF071312.1 | 13 | 1875 | AAC33899.1 | |
| AF087688.1 | AAC36309.1 | |||
| AK010383.1 | 13 | 1912 | BAB26900.1 | |
| AK037757.1 | 13 | 1912 | BAE20520.1 | |
| AK084421.1 | 13 | 1886 | BAC39179.1 | |
| AK132207.1 | 13 | 1882 | BAE21033.1 | |
| AK145225.1 | 12 | 1689 | ||
| AK146153.1 | 13 | 1876 | BAE26937.1 | |
| BC023096.1 | 13 | 1870 | AAH23096.1 |

Genomic Map