Assay Details
Target Gene Details
| Entrez Gene ID: | 12048 | 
| Gene Name: | BCL2-like 1 | 
| Gene Aliases: | Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 | 
| Location: | Chr.2:152758732-152832581 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Bcl2l1 | NM_001289739.1 | NP_001276668.1 | ||
| U51277.1 | AAC53458.1 | 
Target Gene Details
| Entrez Gene ID: | 84682 | 
| Gene Name: | cytochrome c oxidase subunit IV isoform 2 | 
| Gene Aliases: | Cox4b, CoxIV-2 | 
| Location: | Chr.2:152754173-152765037 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Cox4i2 | NM_053091.2 | 5 | 479 | NP_444321.1 | 
| AF271382.1 | 5 | 479 | AAK49433.1 | |
| BC049623.1 | 4 | 488 | AAH49623.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top