Assay Details
Target Gene Details
Entrez Gene ID: | 67136 |
Gene Name: | kelch repeat and BTB (POZ) domain containing 4 |
Gene Aliases: |
2510026C23Rik |
Location: |
Chr.2:90904724-90910561 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kbtbd4 | NM_025991.3 | 2 | 336 | NP_080267.1 |
| AK010998.1 | 2 | 337 | BAB27317.1 | |
| AK014373.1 | 2 | 262 | BAB29305.1 | |
| BC025103.1 | 2 | 241 | AAH25103.1 |

Genomic Map