Assay Details
Target Gene Details
Entrez Gene ID: | 381379 |
Gene Name: | mediator complex subunit 19 |
Gene Aliases: |
2410018M14Rik, 3110040A13Rik, LCMR1 |
Location: |
Chr.2:84678402-84688215 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med19 | NM_025885.3 | 5 | 934 | NP_080161.1 |
| AK005399.1 | 5 | 902 | BAC25114.1 | |
| AK010552.1 | 5 | 912 | ||
| AK139430.1 | 1 | 199 | BAE24010.1 | |
| AK160579.1 | 5 | 900 | BAE35886.1 | |
| AK168787.1 | 5 | 902 | BAE40621.1 | |
| BB611514.1 | 5 | 934 | ||
| BC025475.1 | 4 | 508 | AAH25475.1 | |
| BC044207.1 | 5 | 796 | AAH44207.1 | |
| BC082559.1 | 5 | 899 | AAH82559.1 |

Genomic Map