Assay Details
Target Gene Details
Entrez Gene ID: | 74450 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
4933409I19Rik, AI642621 |
Location: |
Chr.2:131262011-131299188 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pank2 | NM_153501.2 | 3 | 871 | NP_705721.2 |
| AK016755.1 | ||||
| AK082821.1 | 3 | 877 | ||
| AK154023.1 | ||||
| AK154080.1 | 3 | 835 | BAE32361.1 | |
| AK159175.1 | 3 | 862 | ||
| AK167861.1 | 3 | 579 | BAE39879.1 | |
| AK180859.1 | ||||
| AK186345.1 | ||||
| AK208629.1 | ||||
| AK211080.1 | ||||
| BC106184.1 | 3 | 825 | AAI06185.1 |
More Information
Set Membership: |
|

Genomic Map