Assay Details
Target Gene Details
Entrez Gene ID: | 67578 |
Gene Name: | protein associated with topoisomerase II homolog 2 (yeast) |
Gene Aliases: |
4930424G05Rik, Pat1a |
Location: |
Chr.2:122120103-122186189 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Patl2 | NM_026251.2 | 18 | 2210 | NP_080527.2 |
| AK010811.1 | 11 | 1278 | ||
| AK015195.1 | 18 | 2214 | BAB29742.1 |
More Information
Set Membership: |
|

Genomic Map