Assay Details
Target Gene Details
Entrez Gene ID: | 208263 |
Gene Name: | torsin A interacting protein 1 |
Gene Aliases: |
LAP1, Lap1B |
Location: |
Chr.1:156004599-156036480 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip1 | NM_001160018.1 | 10 | 3784 | NP_001153490.1 |
| NM_001160019.1 | 10 | 3198 | NP_001153491.1 | |
| NM_144791.2 | 7 | 3616 | NP_659040.2 | |
| AK079026.1 | 1 | 1987 |

Genomic Map