Assay Details
Target Gene Details
Entrez Gene ID: | 110078 |
Gene Name: | brain glycogen phosphorylase |
Gene Aliases: |
- |
Location: |
Chr.2:150786796-150831748 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pygb | NM_153781.1 | 1 | 102 | NP_722476.1 |
| AK032769.1 | 1 | 115 | BAE20482.1 | |
| AK134667.1 | 1 | 112 | BAE22233.1 | |
| AK142716.1 | 1 | 111 | ||
| AK147763.1 | 1 | 134 | BAE28122.1 | |
| AK168381.1 | 1 | 112 | ||
| AK169052.1 | 1 | 137 | BAE40840.1 | |
| AK169631.1 | 1 | 115 | ||
| BC035283.1 | 1 | 102 | AAH35283.1 |
More Information
Set Membership: |
|

Genomic Map