Assay Details
Target Gene Details
Entrez Gene ID: | 68365 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
0610030G24Rik, 2810475J17Rik, A830021G03Rik, AI314285, AI649155, D030017L14Rik |
Location: |
Chr.2:35180205-35201431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab14 | NM_026697.3 | 1 | 303 | NP_080973.1 |
AJ968955.1 | 1 | 158 | CAI92125.1 | |
AJ968956.1 | 1 | 158 | CAI92126.1 | |
AJ968957.1 | 1 | 158 | CAI92127.1 | |
AJ968958.1 | 1 | 158 | CAI92128.1 | |
AJ968959.1 | 1 | 158 | CAI92129.1 | |
AJ968960.1 | 1 | 158 | CAI92130.1 | |
AK002704.1 | 1 | 175 | BAB22298.1 | |
AK013410.1 | 1 | 156 | ||
AK043699.1 | 1 | 182 | ||
AK133963.1 | 1 | 203 | BAE21957.1 | |
AK135751.1 | 1 | 192 | ||
AK160859.1 | 1 | 303 | BAE36051.1 | |
AK165812.1 | 1 | 150 | BAE38391.1 | |
AK168551.1 | 1 | 160 | BAE40426.1 | |
AK168867.1 | 1 | 172 | BAE40686.1 | |
AK169396.1 | 1 | 174 | BAE41143.1 | |
AK169946.1 | 1 | 165 | ||
BC009085.1 | AAH09085.1 | |||
BC025139.1 | 1 | 174 | AAH25139.1 | |
BC056648.1 | 1 | 176 | AAH56648.1 |
