Assay Details
Target Gene Details
Entrez Gene ID: | 68365 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
0610030G24Rik, 2810475J17Rik, A830021G03Rik, AI314285, AI649155, D030017L14Rik |
Location: |
Chr.2:35180205-35201431 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab14 | NM_026697.3 | 8 | 2710 | NP_080973.1 |
AK133963.1 | 8 | 2610 | BAE21957.1 | |
AK151834.1 | 1 | 1744 | BAE30727.1 | |
AK160859.1 | 8 | 2710 | BAE36051.1 | |
AK166863.1 | 1 | 2729 | ||
AK168551.1 | 8 | 2567 | BAE40426.1 | |
AK168867.1 | BAE40686.1 | |||
AK169396.1 | 8 | 2581 | BAE41143.1 | |
BU702057.1 | 1 | 237 |
