Assay Details
Target Gene Details
Entrez Gene ID: | 228998 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 1 |
Gene Aliases: |
AI115377, Arf1gap |
Location: |
Chr.2:180967225-180982524 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap1 | NM_001177706.1 | 12 | 1837 | NP_001171177.1 |
| NM_001177707.1 | 13 | 1714 | NP_001171178.1 | |
| NM_001177708.1 | 12 | 1433 | NP_001171179.1 | |
| NM_001177709.1 | 12 | 1427 | NP_001171180.1 | |
| NM_001177710.1 | 12 | 1383 | NP_001171181.1 | |
| NM_145760.3 | 14 | 1706 | NP_665703.2 | |
| AK030295.1 | 14 | 1662 | BAC26883.1 | |
| AK030520.1 | 12 | 1839 | BAC27002.1 | |
| AK134402.1 | 13 | 1656 | BAE22128.1 | |
| AK145879.1 | 14 | 1690 | BAE26720.1 | |
| AK168007.1 | 14 | 1687 | BAE39994.1 | |
| AK168602.1 | 12 | 1414 | BAE40469.1 | |
| AK197308.1 | ||||
| AK201622.1 | ||||
| BC016228.1 | 12 | 1341 | ||
| BC052922.1 | 12 | 1376 | ||
| BC059817.1 | 12 | 1366 | AAH59817.1 |
More Information
Set Membership: |
|

Genomic Map