Assay Details
Target Gene Details
Entrez Gene ID: | 77771 |
Gene Name: | cysteine-serine-rich nuclear protein 3 |
Gene Aliases: |
A330102K23Rik, CSRNP-3, Csnrp3, Mbu1, taip-2 |
Location: |
Chr.2:65845767-66031546 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csrnp3 | NM_001290665.1 | 4 | 801 | NP_001277594.1 |
NM_001290666.1 | 4 | 423 | NP_001277595.1 | |
NM_153409.5 | 6 | 845 | NP_700458.3 | |
NM_178634.2 | 5 | 683 | NP_848749.2 | |
AB091688.1 | 3 | 487 | BAC16315.1 | |
AK039150.1 | 6 | 771 | BAC30257.1 | |
AK082649.1 | 5 | 608 | BAC38559.1 | |
AK158873.1 | 4 | 803 | BAE34706.1 | |
BC055701.1 | 4 | 335 | AAH55701.1 | |
EF210820.1 | 6 | 817 |
