Assay Details
Target Gene Details
Entrez Gene ID: | 14347 |
Gene Name: | fucosyltransferase 7 |
Gene Aliases: |
AI853193, FTVII, Fuc-TVII, FucT-VII |
Location: |
Chr.2:25422351-25426374 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fut7 | NM_001177366.1 | NP_001170837.1 | ||
| NM_001177367.1 | NP_001170838.1 | |||
| NM_001289453.1 | NP_001276382.1 | |||
| NM_001289454.1 | NP_001276383.1 | |||
| NM_001289455.1 | NP_001276384.1 | |||
| NM_001289456.1 | NP_001276385.1 | |||
| NM_013524.3 | NP_038552.1 | |||
| AK154486.1 | ||||
| GQ424189.1 | ||||
| GQ424190.1 | ||||
| GQ424191.1 | ||||
| GQ424192.1 | 1 | 271 |
More Information
Set Membership: |
|

Genomic Map