Assay Details
Target Gene Details
Entrez Gene ID: | 18573 |
Gene Name: | phosphodiesterase 1A, calmodulin-dependent |
Gene Aliases: |
AI987702, AW125737 |
Location: |
Chr.2:79834453-80196518 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde1a | NM_001159582.1 | 1 | 225 | NP_001153054.1 |
| AK021279.1 | 1 | 273 | BAB32359.1 | |
| AK050004.1 | 1 | 86 | ||
| CF741268.1 | 1 | 225 |
More Information
Set Membership: |
|

Genomic Map