Assay Details
Target Gene Details
Entrez Gene ID: | 74189 |
Gene Name: | phosphatase and actin regulator 3 |
Gene Aliases: |
1500003N10Rik, 4930415A02Rik, H17739, Scapin1, Scapinin, mKIAA4224 |
Location: |
Chr.2:178118975-178338492 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr3 | NM_001007154.3 | 13 | 2472 | NP_001007155.1 |
| NM_001177789.1 | 13 | 2690 | NP_001171260.1 | |
| NM_001177790.1 | 13 | 2687 | NP_001171261.1 | |
| NM_001177791.1 | 13 | 2524 | NP_001171262.1 | |
| NM_028806.2 | 13 | 2929 | NP_083082.1 | |
| AJ632220.1 | 13 | 2487 | CAG14990.1 | |
| AJ632221.1 | 13 | 2525 | CAG14991.1 | |
| AJ632222.1 | 13 | 2472 | CAG14992.1 | |
| AK005136.1 | 13 | 2494 | BAB23834.1 | |
| AK220554.1 | 13 | 2687 | BAD90539.1 |
More Information
Set Membership: |
|

Genomic Map