Assay Details
Target Gene Details
Entrez Gene ID: | 78774 |
Gene Name: | cilia and flagella associated protein 61 |
Gene Aliases: |
4930529M08Rik, BB178539, BC024760 |
Location: |
Chr.2:145934784-145964226 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfap61 | NM_175280.3 | 7 | 1406 | NP_780489.2 |
| AK038772.1 | 7 | 1407 |
More Information
Set Membership: |
|

Genomic Map