Assay Details
Target Gene Details
Entrez Gene ID: | 14683 |
Gene Name: | GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus |
Gene Aliases: |
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH |
Location: |
Chr.2:174283444-174346744 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnas | NM_019690.2 | 2 | 334 | NP_062664.2 |
| NM_022000.2 | 2 | 334 | NP_068840.2 | |
| AF107847.1 | 2 | 329 | AAD11805.1 | |
| AF107848.1 | 2 | 332 | AAD11806.1 | |
| AF175305.1 | 2 | 309 | AAD55061.1 | |
| AJ010163.1 | 2 | 300 | CAA09026.1 | |
| AK039035.1 | 2 | 334 | BAC30214.1 | |
| AK161197.1 | 1 | 409 |
Target Gene Details
Entrez Gene ID: | 56802 |
Gene Name: | neuroendocrine secretory protein antisense |
Gene Aliases: |
Gnas-as, Nespos |
Location: |
Chr.2:174281237-174295436 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nespas | NR_002846.2 | |||
| AF173359.1 | 1 | 689 | ||
| AF319945.1 | ||||
| AK153813.1 |
More Information
Set Membership: |
|

Genomic Map