Assay Details
Target Gene Details
Entrez Gene ID: | 18286 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
AI848335, MMTEST29 |
Location: |
Chr.2:29889720-29931746 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Odf2 | NM_001113213.1 | 21 | 2838 | NP_001106684.1 |
| NM_001177659.1 | 21 | 2841 | NP_001171130.1 | |
| AK143726.1 | 21 | 2745 | BAE25517.1 | |
| AK165588.1 | 21 | 2745 | ||
| BC057001.1 | 22 | 2895 | AAH57001.1 |

Genomic Map