Assay Details
Target Gene Details
Entrez Gene ID: | 66766 |
Gene Name: | transmembrane 239 |
Gene Aliases: |
4933425O20Rik |
Location: |
Chr.2:130406522-130407794 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem239 | NM_025753.3 | 2 | 955 | NP_080029.1 |
| AK016923.1 | 2 | 970 | BAC25500.1 | |
| BC049728.1 | 2 | 956 | AAH49728.1 |

Genomic Map