Assay Details
Target Gene Details
Entrez Gene ID: | 77683 |
Gene Name: | euchromatic histone methyltransferase 1 |
Gene Aliases: |
9230102N17Rik, D330003E03, Eu-HMTase1, GLP, GLP1, KMT1D, mKIAA1876 |
Location: |
Chr.2:24790769-24919643 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ehmt1 | NM_001012518.3 | 27 | 3975 | NP_001012536.2 |
| NM_001109686.2 | 26 | 3816 | NP_001103156.1 | |
| NM_001109687.2 | 26 | 3831 | NP_001103157.1 | |
| NM_172545.4 | 27 | 3954 | NP_766133.2 | |
| AB205007.1 | 27 | 3981 | BAD90007.1 | |
| AK049454.1 | 24 | 3225 | BAC33756.1 | |
| AK052174.1 | 27 | 3938 | BAC34869.1 | |
| AK082062.1 | 27 | 3965 | BAC38402.1 | |
| AK129461.1 | BAC98271.1 | |||
| AK149125.1 | 7 | 898 | BAE28744.1 | |
| BC056938.1 | 24 | 3532 | AAH56938.1 | |
| BC089302.1 | 24 | 3684 | AAH89302.1 |

Genomic Map