Assay Details
Target Gene Details
Entrez Gene ID: | 67996 |
Gene Name: | serine/arginine-rich splicing factor 6 |
Gene Aliases: |
1210001E11Rik, AI314910, AW146126, Sfrs6 |
Location: |
Chr.2:162931508-162937121 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf6 | NM_026499.4 | 6 | 1730 | NP_080775.3 |
| AK004831.1 | BAB23599.1 | |||
| AK011561.1 | ||||
| AK012605.1 | ||||
| AK051608.1 | BAC34689.1 | |||
| AK133872.1 | 6 | 1711 | BAE21902.1 | |
| AK146293.1 | 6 | 1708 | BAE27050.1 | |
| AK159519.1 | 6 | 1708 | BAE35148.1 | |
| BC012039.1 | 6 | 1717 | AAH12039.1 |
More Information
Set Membership: |
|

Genomic Map