Assay Details
Target Gene Details
Entrez Gene ID: | 76161 |
Gene Name: | lysosomal-associated membrane protein family, member 5 |
Gene Aliases: |
3110035N03Rik, 6330527O06Rik, BAD-LAMP, LAMP-5 |
Location: |
Chr.2:136051959-136069917 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamp5 | NM_029530.2 | 6 | 1796 | NP_083806.2 |
| AI839765.1 | 1 | 177 | ||
| AK014127.1 | 4 | 1238 | BAB29169.1 | |
| AK018222.1 | 6 | 1494 | ||
| BC004791.1 | 4 | 1072 | AAH04791.1 | |
| BU706723.1 | 1 | 197 |
More Information
Set Membership: |
|

Genomic Map