Assay Details
Target Gene Details
Entrez Gene ID: | 69270 |
Gene Name: | GINS complex subunit 1 (Psf1 homolog) |
Gene Aliases: |
2810418N01Rik, Gins4, PSF1, mKIAA0186 |
Location: |
Chr.2:150909557-150931280 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gins1 | NM_027014.1 | 7 | 720 | NP_081290.1 |
| AK013116.1 | 7 | 713 | BAB28658.1 | |
| AK129081.1 | 6 | 511 | BAC97891.1 |
More Information
Set Membership: |
|

Genomic Map