Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
