Assay Details
Target Gene Details
Entrez Gene ID: | 17865 |
Gene Name: | myeloblastosis oncogene-like 2 |
Gene Aliases: |
B-Myb, Bmyb |
Location: |
Chr.2:163054333-163084688 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mybl2 | NM_008652.2 | 14 | 2685 | NP_032678.1 |
AK028497.1 | 14 | 2685 | BAC25979.1 | |
AK207983.1 | ||||
BC050842.1 | AAH50842.1 | |||
X70472.1 | CAA49898.1 |
