Assay Details
Target Gene Details
Entrez Gene ID: | 170791 |
Gene Name: | RNA binding motif protein 39 |
Gene Aliases: |
1500012C14Rik, 2310040E03Rik, B330012G18Rik, C79248, R75070, Rnpc2, caper |
Location: |
Chr.2:156147239-156180238 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm39 | NM_001291114.1 | 1 | 170 | NP_001278043.1 |
| NM_133242.3 | 1 | 170 | NP_573505.2 | |
| AK005227.1 | 1 | 119 | ||
| AK005234.1 | 1 | 119 | ||
| AK009719.1 | 1 | 83 | ||
| AK147076.1 | 1 | 115 | BAE27657.1 | |
| AK154989.1 | 1 | 101 | BAE32977.1 | |
| AK155976.1 | 1 | 119 | ||
| AK156737.1 | 1 | 79 | ||
| AK167355.1 | 1 | 115 | BAE39454.1 | |
| BC086645.1 | 1 | 93 | AAH86645.1 |
More Information
Set Membership: |
|

Genomic Map