Assay Details
Target Gene Details
Entrez Gene ID: | 12010 |
Gene Name: | beta-2 microglobulin |
Gene Aliases: |
Ly-m11, beta2-m, beta2m |
Location: |
Chr.2:122147687-122153082 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B2m | NM_009735.3 | 2 | 305 | NP_033865.2 |
| AK019389.1 | 2 | 308 | BAB31695.1 | |
| AK020632.1 | 2 | 307 | BAB32158.1 | |
| AK028378.1 | 2 | 307 | BAC25917.1 | |
| AK153255.1 | 2 | 307 | BAE31846.1 | |
| AK166067.1 | 2 | 308 | BAE38551.1 | |
| BC085164.1 | 2 | 260 | AAH85164.1 | |
| M10416.1 | AAA39537.1 | |||
| X01838.1 | 2 | 306 | CAA25970.1 |
Target Gene Details
Entrez Gene ID: | 67578 |
Gene Name: | protein associated with topoisomerase II homolog 2 (yeast) |
Gene Aliases: |
4930424G05Rik, Pat1a |
Location: |
Chr.2:122120103-122186189 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Patl2 | NM_026251.2 | NP_080527.2 | ||
| AK015195.1 | BAB29742.1 | |||
| BY714747.1 |
More Information
Set Membership: |
|

Genomic Map