Assay Details
Target Gene Details
Entrez Gene ID: | 12010 |
Gene Name: | beta-2 microglobulin |
Gene Aliases: |
Ly-m11, beta2-m, beta2m |
Location: |
Chr.2:122147687-122153082 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B2m | NM_009735.3 | 4 | 788 | NP_033865.2 |
| AK019389.1 | 4 | 791 | BAB31695.1 | |
| AK020632.1 | 4 | 790 | BAB32158.1 | |
| AK153255.1 | 4 | 790 | BAE31846.1 | |
| AK166067.1 | 4 | 791 | BAE38551.1 |
Target Gene Details
Entrez Gene ID: | 67578 |
Gene Name: | protein associated with topoisomerase II homolog 2 (yeast) |
Gene Aliases: |
4930424G05Rik, Pat1a |
Location: |
Chr.2:122120103-122186189 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Patl2 | NM_026251.2 | NP_080527.2 | ||
| AK015195.1 | BAB29742.1 | |||
| BY714747.1 |
More Information
Set Membership: |
|

Genomic Map