Assay Details
Target Gene Details
Entrez Gene ID: | 18795 |
Gene Name: | phospholipase C, beta 1 |
Gene Aliases: |
3110043I21Rik, AI132408, Plcb, mKIAA0581 |
Location: |
Chr.2:134786164-135475258 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcb1 | NM_001145830.1 | 1 | 232 | NP_001139302.1 |
| NM_019677.2 | 1 | 232 | NP_062651.2 | |
| AK040911.1 | 1 | 223 | ||
| AK141164.1 | 1 | 179 | ||
| AK141464.1 | 1 | 168 | ||
| AK143153.1 | 1 | 233 | BAE25284.1 | |
| BC058710.1 | 1 | 199 | AAH58710.1 |
More Information
Set Membership: |
|

Genomic Map