Assay Details
Target Gene Details
Entrez Gene ID: | 56878 |
Gene Name: | RNA binding motif, single stranded interacting protein 1 |
Gene Aliases: |
2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1 |
Location: |
Chr.2:60750193-60963450 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbms1 | NM_020296.2 | 1 | 431 | NP_064692.2 |
AB026569.1 | 1 | 153 | BAA77262.1 | |
AK135280.1 | 1 | 118 | BAE22471.1 | |
AK147109.1 | 1 | 432 | BAE27681.1 | |
AK184927.1 | 1 | 75 | ||
BC016501.1 | 1 | 419 |
