Assay Details
Target Gene Details
Entrez Gene ID: | 56878 |
Gene Name: | RNA binding motif, single stranded interacting protein 1 |
Gene Aliases: |
2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1 |
Location: |
Chr.2:60750193-60963450 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbms1 | NM_001141931.1 | 14 | 1983 | NP_001135403.1 |
NM_001141932.1 | 15 | 2031 | NP_001135404.1 | |
NM_020296.2 | 14 | 2176 | NP_064692.2 | |
AA175245.1 | 1 | 374 | ||
AK147109.1 | 14 | 2177 | BAE27681.1 | |
AK159851.1 | 2 | 584 | BAE35428.1 | |
AK161102.1 | 15 | 1981 | BAE36195.1 | |
AK161416.1 | 10 | 1313 | BAE36384.1 |
