Assay Details
Target Gene Details
Entrez Gene ID: | 20612 |
Gene Name: | sialic acid binding Ig-like lectin 1, sialoadhesin |
Gene Aliases: |
Cd169, Siglec-1, Sn |
Location: |
Chr.2:131069220-131086787 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Siglec1 | NM_011426.3 | 6 | 2151 | NP_035556.3 |
BC141335.1 | 6 | 1745 | ||
BC141336.1 | 6 | 1741 | ||
Z36233.1 | 6 | 2149 | CAA85268.1 | |
Z36293.1 | 6 | 2149 | CAA85290.1 |
