Assay Details
Target Gene Details
Entrez Gene ID: | 67388 |
Gene Name: | RIKEN cDNA 1110008F13 gene |
Gene Aliases: |
AI316789 |
Location: |
Chr.2:156863122-156873563 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008F13Rik | NM_026124.3 | 4 | 684 | NP_080400.1 |
| AK002452.1 | 4 | 642 | BAB22111.1 | |
| AK003568.1 | 4 | 679 | BAB22864.1 | |
| AK078178.1 | 4 | 679 | ||
| AK184175.1 | ||||
| AK189929.1 | ||||
| AW493966.1 | 1 | 382 | ||
| BC019445.1 | 4 | 622 | AAH19445.1 |
Target Gene Details
Entrez Gene ID: | 77799 |
Gene Name: | Src-like-adaptor 2 |
Gene Aliases: |
A930009E21Rik, AI430952, SLAP-2, SLAP2 |
Location: |
Chr.2:156872922-156887241 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sla2 | NM_029983.5 | 8 | 2234 | NP_084259.1 |
| AK030877.1 | 8 | 2235 | BAC27168.1 | |
| AK088672.1 | 8 | 2361 | BAC40495.1 | |
| AK138709.1 | 8 | 2235 | BAE23754.1 |
More Information
Set Membership: |
|

Genomic Map