Assay Details
Target Gene Details
Entrez Gene ID: | 407823 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2B |
Gene Aliases: |
5830435C13Rik, BC053917, D2Ertd794e |
Location: |
Chr.2:59899363-60209846 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baz2b | NM_001001182.3 | 28 | 4986 | NP_001001182.2 |
| AK143439.1 | 1 | 123 | ||
| BC150814.1 | 30 | 4995 | ||
| CA319831.1 | 2 | 435 |
More Information
Set Membership: |
|

Genomic Map